COM448656240 Combi-Blocks, Inc.

4,5,6,7-Tetrahydro-2h-pyrazolo[4,3-c]pyridine

  • MDL number MFCD13176187
  • Cas Number : 933742-87-1
  • Molecular Weight : 123.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 98
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2713
Availability details for
Quantity
  • Availability
  • Add to Cart