COM448656304 Combi-Blocks, Inc.

3-Amino-5-fluorobenzamide

  • MDL number MFCD08447276
  • Cas Number : 1036757-40-0
  • Molecular Weight : 154.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2781

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2781
Availability details for
Quantity
  • Availability
  • Add to Cart