COM448656315 Combi-Blocks, Inc.

2'-Deoxy-n2-dimethylaminomethylene-guanosine

  • MDL number MFCD08704073
  • Cas Number : 17331-13-4
  • Molecular Weight : 322.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2793
Availability details for
Quantity
  • Availability
  • Add to Cart