COM448656319 Combi-Blocks, Inc.

Meso-octamethylcalix(4)pyrrole

  • MDL number MFCD00208353
  • Cas Number : 4475-42-7
  • Molecular Weight : 428.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2797

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2797
Availability details for
Quantity
  • Availability
  • Add to Cart