COM448656400 Combi-Blocks, Inc.

4-Bromo-5-methoxy-1h-indazole

  • MDL number MFCD17676852
  • Cas Number : 1192004-62-8
  • Molecular Weight : 227.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2879
Availability details for
Quantity
  • Availability
  • Add to Cart