COM448656485 Combi-Blocks, Inc.

L-Allose

  • MDL number MFCD00085373
  • Cas Number : 7635-11-2
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2971
Availability details for
Quantity
  • Availability
  • Add to Cart