COM448656500 Combi-Blocks, Inc.

1,2,2,2-Tetrafluoroethylsulfonyl fluoride

  • MDL number MFCD00042529
  • Cas Number : 2127-74-4
  • Molecular Weight : 184.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2988
Availability details for
Quantity
  • Availability
  • Add to Cart