COM448656548 Combi-Blocks, Inc.

7-Bromo-1-isopropyl-3,4-dihydroisoquinoline

  • MDL number MFCD09909334
  • Cas Number : 371220-28-9
  • Molecular Weight : 252.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3041

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3041
Availability details for
Quantity
  • Availability
  • Add to Cart