COM448656558 Combi-Blocks, Inc.

Bis propargyl-PEG6

  • MDL number MFCD10699797
  • Cas Number : 400775-35-1
  • Molecular Weight : 358.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3053

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3053
Availability details for
Quantity
  • Availability
  • Add to Cart