COM448656567 Combi-Blocks, Inc.

2-(2-Hydroxyethylamino)-n-(4-chlorophenyl)acetamide

  • MDL number MFCD09037910
  • Cas Number : 215649-69-7
  • Molecular Weight : 228.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3063
Availability details for
Quantity
  • Availability
  • Add to Cart