COM448656623 Combi-Blocks, Inc.

Di-p-tolylphosphine

  • MDL number MFCD01630843
  • Cas Number : 1017-60-3
  • Molecular Weight : 214.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3125

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3125
Availability details for
Quantity
  • Availability
  • Add to Cart