COM448656638 Combi-Blocks, Inc.

Bis(sulphosuccinimidyl)suberate

  • MDL number MFCD01861948
  • Cas Number : 82436-77-9
  • Molecular Weight : 528.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3140
Availability details for
Quantity
  • Availability
  • Add to Cart