COM448656651 Combi-Blocks, Inc.

2-Amino-3,5-difluorobenzonitrile

  • MDL number MFCD18826239
  • Cas Number : 126674-94-0
  • Molecular Weight : 154.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3153

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3153
Availability details for
Quantity
  • Availability
  • Add to Cart