COM448656758 Combi-Blocks, Inc.

1,1'-Iminodianthraquinone

  • MDL number MFCD00019132
  • Cas Number : 82-22-4
  • Molecular Weight : 429.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3266

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3266
Availability details for
Quantity
  • Availability
  • Add to Cart