COM448656791 Combi-Blocks, Inc.

1-(3-Methyl-benzoyl)-piperazine

  • MDL number MFCD04116565
  • Cas Number : 100939-91-1
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3299

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3299
Availability details for
Quantity
  • Availability
  • Add to Cart