COM448656862 Combi-Blocks, Inc.

Ethyl 2,3-di-o-benzoyl-4,6-o-benzylidene-beta-d-thiogalactopyranoside

  • MDL number MFCD13182931
  • Cas Number : 56119-30-3
  • Molecular Weight : 520.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3372
Availability details for
Quantity
  • Availability
  • Add to Cart