COM448656873 Combi-Blocks, Inc.

(Perfluoro-n-propyl)phenyliodonium trifluoromethanesulfonate

  • MDL number MFCD00135121
  • Cas Number : 77758-79-3
  • Molecular Weight : 522.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3383

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3383
Availability details for
Quantity
  • Availability
  • Add to Cart