COM448656880 Combi-Blocks, Inc.

tert-Butyl 1-amino-6-azaspiro[3.4]octane-6-carboxylate

  • MDL number MFCD17016201
  • Cas Number : 1251010-30-6
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3390

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3390
Availability details for
Quantity
  • Availability
  • Add to Cart