COM448656929 Combi-Blocks, Inc.

Bis(1h,1h,2h,2h-perfluorodecyl)disulfide

  • MDL number MFCD08457554
  • Cas Number : 42977-21-9
  • Molecular Weight : 958.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3445

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3445
Availability details for
Quantity
  • Availability
  • Add to Cart