COM448656981 Combi-Blocks, Inc.

Mal-dpeg(4)-cooh

  • MDL number MFCD13184954
  • Cas Number : 1263045-16-4
  • Molecular Weight : 416.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3498
Availability details for
Quantity
  • Availability
  • Add to Cart