COM448656990 Combi-Blocks, Inc.

(4-Methyl-2-phenyl-1,3-oxazol-5-ylmethyl)amine

  • MDL number MFCD11109320
  • Cas Number : 165735-97-7
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3509
Availability details for
Quantity
  • Availability
  • Add to Cart