COM448657059 Combi-Blocks, Inc.

(Z)-Methyl 2-(tert-butoxycarbonylamino)but-2-enoate

  • MDL number MFCD15142810
  • Cas Number : 63658-16-2
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3583

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3583
Availability details for
Quantity
  • Availability
  • Add to Cart