COM448657200 Combi-Blocks, Inc.

Arachidic anhydride

  • MDL number MFCD00056235
  • Cas Number : 55726-22-2
  • Molecular Weight : 607.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3736

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3736
Availability details for
Quantity
  • Availability
  • Add to Cart