COM448657209 Combi-Blocks, Inc.

Neopentyl glycol mono(hydroxypivalate)

  • MDL number MFCD00059597
  • Cas Number : 1115-20-4
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3745

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3745
Availability details for
Quantity
  • Availability
  • Add to Cart