COM448657278 Combi-Blocks, Inc.

5-[3,5-Bis(trifluoromethyl)phenyl]-2,4-dihydro-3h-1,2,4-triazole-3-thione

  • MDL number MFCD00177275
  • Cas Number : 175276-77-4
  • Molecular Weight : 313.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3817

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3817
Availability details for
Quantity
  • Availability
  • Add to Cart