COM448657300 Combi-Blocks, Inc.

Ethyl 4,6-o-benzylidene-2-o-levulinoyl-beta-d-thioglucopyranoside

  • MDL number MFCD18643042
  • Molecular Weight : 410.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3840
Availability details for
Quantity
  • Availability
  • Add to Cart