COM448657324 Combi-Blocks, Inc.

2-(2,5-Dibromophenyl)acetonitrile

  • MDL number MFCD11618143
  • Cas Number : 74533-21-4
  • Molecular Weight : 274.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3868

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3868
Availability details for
Quantity
  • Availability
  • Add to Cart