COM448657414 Combi-Blocks, Inc.

2'-Deoxyisoguanosine

  • MDL number MFCD15145285
  • Cas Number : 106449-56-3
  • Molecular Weight : 267.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3966

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3966
Availability details for
Quantity
  • Availability
  • Add to Cart