COM448657429 Combi-Blocks, Inc.

Carbazochrome

  • MDL number MFCD00130253
  • Cas Number : 69-81-8
  • Molecular Weight : 236.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3983

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-3983
Availability details for
Quantity
  • Availability
  • Add to Cart