COM448657464 Combi-Blocks, Inc.

Mntbap

  • MDL number MFCD06227760
  • Cas Number : 55266-18-7
  • Molecular Weight : 879.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4025
Availability details for
Quantity
  • Availability
  • Add to Cart