COM448657482 Combi-Blocks, Inc.

2-(4-Hydroxy-phenyl)-1-oxa-7-aza-spiro[4.5]decane-7-carboxylic acid tert-butyl ester

  • MDL number MFCD14585374
  • Cas Number : 1206970-20-8
  • Molecular Weight : 333.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4045
Availability details for
Quantity
  • Availability
  • Add to Cart