COM448657521 Combi-Blocks, Inc.

1-Oxa-4,8-diazaspiro[5.5]undecane-8-carboxylic acid tert-butyl ester

  • MDL number MFCD11227062
  • Cas Number : 1160247-05-1
  • Molecular Weight : 256.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4086

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4086
Availability details for
Quantity
  • Availability
  • Add to Cart