COM448657542 Combi-Blocks, Inc.

[N-[1-[2-(2-Pyridylcarboxamido)phenyl]ethylidene]glycinato]nickel

  • MDL number MFCD01823363
  • Cas Number : 264921-97-3
  • Molecular Weight : 354.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4108
Availability details for
Quantity
  • Availability
  • Add to Cart