COM448657562 Combi-Blocks, Inc.

Ethyl 5-(pyridin-2-yl)-1h-pyrazole-3-carboxylate

  • MDL number MFCD13190394
  • Cas Number : 174139-65-2
  • Molecular Weight : 217.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4128

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4128
Availability details for
Quantity
  • Availability
  • Add to Cart