COM448657618 Combi-Blocks, Inc.

Isobutyl isovalerate

  • MDL number MFCD00048349
  • Cas Number : 589-59-3
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4185

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4185
Availability details for
Quantity
  • Availability
  • Add to Cart