COM448657638 Combi-Blocks, Inc.

3-Chloro-4-methylbenzenesulfinic acid sodium salt

  • MDL number MFCD03093774
  • Cas Number : 144772-86-1
  • Molecular Weight : 212.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4205
Availability details for
Quantity
  • Availability
  • Add to Cart