COM448657710 Combi-Blocks, Inc.

(4-(3-Chlorophenyl)-1h-imidazol-2-yl)methanamine

  • MDL number MFCD10696694
  • Cas Number : 944897-81-8
  • Molecular Weight : 207.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4280

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4280
Availability details for
Quantity
  • Availability
  • Add to Cart