COM448657742 Combi-Blocks, Inc.

1,2,2,3-Tetrachloropropane

  • MDL number MFCD00039362
  • Cas Number : 13116-53-5
  • Molecular Weight : 181.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4317
Availability details for
Quantity
  • Availability
  • Add to Cart