COM448657760 Combi-Blocks, Inc.

1-Benzyl-1,7-diazaspiro[4.4]nonane

  • MDL number MFCD09056754
  • Cas Number : 128244-01-9
  • Molecular Weight : 216.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4336

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4336
Availability details for
Quantity
  • Availability
  • Add to Cart