COM448657781 Combi-Blocks, Inc.

3-Bromophenylglyoxal hydrate

  • MDL number MFCD08705873
  • Cas Number : 106134-16-1
  • Molecular Weight : 231.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4358

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4358
Availability details for
Quantity
  • Availability
  • Add to Cart