COM448657789 Combi-Blocks, Inc.

Ethyl 4-oxotetrahydrothiophene-3-carboxylate

  • MDL number MFCD01318533
  • Cas Number : 78647-31-1
  • Molecular Weight : 174.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4367

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4367
Availability details for
Quantity
  • Availability
  • Add to Cart