COM448657877 Combi-Blocks, Inc.

6-Bromoquinoline-2-carbaldehyde

  • MDL number MFCD06824164
  • Cas Number : 98948-91-5
  • Molecular Weight : 236.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4460

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4460
Availability details for
Quantity
  • Availability
  • Add to Cart