COM448657910 Combi-Blocks, Inc.

Trans-tert-butyl 3-amino-4-methylpiperidine-1-carboxylate

  • MDL number MFCD18791219
  • Cas Number : 1271810-27-5
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4495

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4495
Availability details for
Quantity
  • Availability
  • Add to Cart