COM448657958 Combi-Blocks, Inc.

3-Amino-2,2-dimethyl-1-propanol

  • MDL number MFCD00059847
  • Cas Number : 26734-09-8
  • Molecular Weight : 103.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4546

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4546
Availability details for
Quantity
  • Availability
  • Add to Cart