COM448657975 Combi-Blocks, Inc.

Vanadium oxytrifluoride

  • MDL number MFCD00042555
  • Cas Number : 13709-31-4
  • Molecular Weight : 123.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4564

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4564
Availability details for
Quantity
  • Availability
  • Add to Cart