COM448658020 Combi-Blocks, Inc.

Pth-methionine

  • MDL number MFCD00038433
  • Cas Number : 4370-90-5
  • Molecular Weight : 266.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4610
Availability details for
Quantity
  • Availability
  • Add to Cart