COM448658022 Combi-Blocks, Inc.

Cis-1-benzyl-5-ethoxycarbonylhexahydropyrrolo[3,4-b]pyrrole

  • MDL number MFCD27500635
  • Cas Number : 894853-99-7
  • Molecular Weight : 274.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4612

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4612
Availability details for
Quantity
  • Availability
  • Add to Cart