COM448658193 Combi-Blocks, Inc.

2-Chloro-6,7-difluoro-3-quinolinecarboxaldehyde

  • MDL number MFCD06239276
  • Cas Number : 209909-13-7
  • Molecular Weight : 227.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4793
Availability details for
Quantity
  • Availability
  • Add to Cart