COM448658198 Combi-Blocks, Inc.

3-(2-(Chloroacetamido)ethyl)indole

  • MDL number MFCD00120131
  • Cas Number : 52191-26-1
  • Molecular Weight : 236.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4799

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4799
Availability details for
Quantity
  • Availability
  • Add to Cart