COM448658205 Combi-Blocks, Inc.

H-Thr-gly-gly-oh

  • MDL number MFCD00034321
  • Cas Number : 72748-97-1
  • Molecular Weight : 233.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4808

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4808
Availability details for
Quantity
  • Availability
  • Add to Cart